Assay Details
| Gene Symbol: LOC4326038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785361.2 | XP_015640847.1 | 5 - 6 | 1289 | 94 | |
| GenBank mRNA | AJ437346.1 | - | 4 - 5 | 1200 | 94 | |
| AK063986.1 | - | 5 - 6 | 1267 | 94 | ||
| AK073457.1 | - | 5 - 6 | 1268 | 94 | ||
| AK106560.1 | - | 5 - 6 | 1268 | 94 | ||
| AK122166.1 | - | 5 - 6 | 1298 | 94 | ||
More Information
Set Membership: |
|

LOC4326038
Genomic Map