Assay Details
| Gene Symbol: LOC4327698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766199.1 | XP_015621685.1 | 9 - 10 | 1131 | 126 | |
| XM_015766200.2 | XP_015621686.1 | 9 - 10 | 1067 | 126 | ||
| GenBank mRNA | AK067756.1 | - | 9 - 10 | 1101 | 126 | |
| AK098860.1 | - | 9 - 10 | 1037 | 126 | ||
More Information
Set Membership: |
|

LOC4327698
Genomic Map