Assay Details
Gene Symbol: LOC4327709 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762332.2 | XP_015617818.1 | 13 - 14 | 1790 | 110 | |
XM_015762340.2 | XP_015617826.1 | 13 - 14 | 1742 | 110 | ||
GenBank mRNA | AB638271.1 | - | 13 - 14 | 1515 | 110 | |
AK111583.1 | - | 13 - 14 | 1630 | 110 | ||
AK111762.1 | - | 13 - 14 | 1791 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
