Assay Details
Gene Symbol: LOC4327709 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762332.2 | XP_015617818.1 | 16 - 17 | 2323 | 79 | |
XM_015762340.2 | XP_015617826.1 | 16 - 17 | 2275 | 79 | ||
GenBank mRNA | AB638271.1 | - | 16 - 17 | 2048 | 79 | |
AK111583.1 | - | 16 - 17 | 2163 | 79 | ||
AK111762.1 | - | 16 - 17 | 2324 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
