Assay Details
| Gene Symbol: LOC4327709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762332.2 | XP_015617818.1 | 17 - 18 | 2421 | 98 | |
| XM_015762340.2 | XP_015617826.1 | 17 - 18 | 2373 | 98 | ||
| GenBank mRNA | AB638271.1 | - | 17 - 18 | 2146 | 98 | |
| AK111583.1 | - | 17 - 18 | 2261 | 98 | ||
| AK111762.1 | - | 17 - 18 | 2422 | 98 | ||
More Information
Set Membership: |
|

LOC4327709
Genomic Map