Assay Details
| Gene Symbol: LOC4327709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762332.2 | XP_015617818.1 | 19 - 20 | 2686 | 90 | |
| XM_015762340.2 | XP_015617826.1 | 19 - 20 | 2638 | 90 | ||
| GenBank mRNA | AB638271.1 | - | 19 - 20 | 2411 | 90 | |
| AK111583.1 | - | 19 - 20 | 2526 | 90 | ||
| AK111762.1 | - | 19 - 20 | 2687 | 90 | ||
More Information
Set Membership: |
|

LOC4327709
Genomic Map