Assay Details
Gene Symbol: LOC4327709 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762332.2 | XP_015617818.1 | 21 - 22 | 2991 | 146 | |
XM_015762340.2 | XP_015617826.1 | 21 - 22 | 2943 | 146 | ||
GenBank mRNA | AB638271.1 | - | 21 - 22 | 2716 | 146 | |
AK111583.1 | - | 21 - 22 | 2831 | 146 | ||
AK111762.1 | - | 21 - 22 | 2992 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
