Assay Details
| Gene Symbol: LOC4327709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762332.2 | XP_015617818.1 | 22 - 23 | 3089 | 115 | |
| XM_015762340.2 | XP_015617826.1 | 22 - 23 | 3041 | 115 | ||
| GenBank mRNA | AB638271.1 | - | 22 - 23 | 2814 | 115 | |
| AK111583.1 | - | 22 - 23 | 2929 | 115 | ||
| AK111762.1 | - | 22 - 23 | 3090 | 115 | ||
More Information
Set Membership: |
|

LOC4327709
Genomic Map