Assay Details
Gene Symbol: LOC4327709 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762332.2 | XP_015617818.1 | 6 - 7 | 1109 | 95 | |
GenBank mRNA | AB638271.1 | - | 6 - 7 | 834 | 95 | |
AK111762.1 | - | 6 - 7 | 1110 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
