Assay Details
| Gene Symbol: LOC4327772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766254.1 | XP_015621740.1 | 3 - 4 | 624 | 112 | |
| GenBank mRNA | AK103288.1 | - | 3 - 4 | 627 | 112 | |
More Information
Set Membership: |
|

LOC4327772
Genomic Map