Assay Details
Gene Symbol: LOC4326419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765941.2 | XP_015621427.1 | 10 - 11 | 1267 | 84 | |
GenBank mRNA | AK120431.1 | - | 10 - 11 | 1153 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
