Assay Details
Gene Symbol: LOC4326419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765941.2 | XP_015621427.1 | 9 - 10 | 1159 | 72 | |
GenBank mRNA | AK120431.1 | - | 9 - 10 | 1045 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
