Assay Details
| Gene Symbol: LOC4327920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777524.2 | XP_015633010.1 | 2 - 3 | 434 | 78 | |
| XM_015777533.2 | XP_015633019.1 | 2 - 3 | 251 | 78 | ||
| XM_015777542.2 | XP_015633028.1 | 2 - 3 | 434 | 78 | ||
| XM_015777552.2 | XP_015633038.1 | 2 - 3 | 348 | 78 | ||
| GenBank mRNA | AK072598.1 | - | 2 - 3 | 352 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015777552.2 transcripts.
Set Membership: |
|

LOC4327920
Genomic Map