Assay Details
| Gene Symbol: LOC4327960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770176.2 | XP_015625662.1 | 11 - 12 | 1509 | 80 | |
| GenBank mRNA | AK064082.1 | - | 11 - 12 | 1399 | 80 | |
| AK101426.1 | - | 11 - 12 | 1403 | 80 | ||
More Information
Set Membership: |
|

LOC4327960
Genomic Map