Assay Details
Gene Symbol: LOC4327960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770176.2 | XP_015625662.1 | 3 - 4 | 420 | 92 | |
GenBank mRNA | AK064082.1 | - | 3 - 4 | 310 | 92 | |
AK101426.1 | - | 3 - 4 | 314 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
