Assay Details
Gene Symbol: LOC4327229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766472.2 | XP_015621958.1 | 10 - 11 | 1465 | 131 | |
GenBank mRNA | AB117999.1 | - | 4 - 5 | 661 | 131 | |
AK067886.1 | - | 10 - 11 | 1446 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
