Assay Details
Gene Symbol: LOC4327229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766472.2 | XP_015621958.1 | 2 - 3 | 434 | 72 | |
GenBank mRNA | AK067886.1 | - | 2 - 3 | 415 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
