Assay Details
Gene Symbol: LOC4327229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766472.2 | XP_015621958.1 | 5 - 6 | 687 | 132 | |
GenBank mRNA | AK058422.1 | - | 1 - 2 | 39 | 132 | |
AK067886.1 | - | 5 - 6 | 668 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
