Assay Details
Gene Symbol: LOC4327229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766472.2 | XP_015621958.1 | 6 - 7 | 766 | 86 | |
GenBank mRNA | AK058422.1 | - | 2 - 3 | 118 | 86 | |
AK067886.1 | - | 6 - 7 | 747 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
