Assay Details
Gene Symbol: LOC4327229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766472.2 | XP_015621958.1 | 8 - 9 | 1344 | 64 | |
GenBank mRNA | AB117999.1 | - | 2 - 3 | 540 | 64 | |
AK067886.1 | - | 8 - 9 | 1325 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
