Assay Details
Gene Symbol: LOC4327229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766472.2 | XP_015621958.1 | 9 - 10 | 1442 | 117 | |
GenBank mRNA | AB117999.1 | - | 3 - 4 | 638 | 117 | |
AK067886.1 | - | 9 - 10 | 1423 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
