Assay Details
Gene Symbol: LOC4328009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770572.2 | XP_015626058.1 | 11 - 12 | 3963 | 105 | |
GenBank mRNA | AK106550.1 | - | 2 - 3 | 155 | 105 | |
AK122164.1 | - | 10 - 11 | 4008 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
