Assay Details
Gene Symbol: LOC4328009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770572.2 | XP_015626058.1 | 12 - 13 | 4259 | 81 | |
GenBank mRNA | AK106550.1 | - | 3 - 4 | 451 | 81 | |
AK122164.1 | - | 11 - 12 | 4304 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
