Assay Details
Gene Symbol: LOC4328009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770572.2 | XP_015626058.1 | 2 - 3 | 2616 | 107 | |
GenBank mRNA | AK122164.1 | - | 2 - 3 | 2579 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
