Assay Details
Gene Symbol: LOC4328009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770572.2 | XP_015626058.1 | 6 - 7 | 3156 | 132 | |
GenBank mRNA | AK122164.1 | - | 5 - 6 | 3201 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
