Assay Details
| Gene Symbol: LOC4328088 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772149.2 | XP_015627635.1 | 11 - 12 | 1324 | 68 | |
| GenBank mRNA | AK061815.1 | - | 11 - 12 | 1220 | 68 | |
| AK102022.1 | - | 11 - 12 | 1218 | 68 | ||
More Information
Set Membership: |
|

LOC4328088
Genomic Map