Assay Details
| Gene Symbol: LOC4328504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770832.2 | XP_015626318.1 | 3 - 4 | 367 | 98 | |
| GenBank mRNA | AK067511.1 | - | 3 - 4 | 375 | 98 | |
More Information
Set Membership: |
|

LOC4328504
Genomic Map