Assay Details
| Gene Symbol: LOC4328504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770832.2 | XP_015626318.1 | 16 - 17 | 2598 | 103 | |
| XM_015770834.2 | XP_015626320.1 | 15 - 16 | 2552 | 103 | ||
| GenBank mRNA | AK067511.1 | - | 8 - 9 | 3062 | 103 | |
More Information
Set Membership: |
|

LOC4328504
Genomic Map