Assay Details
Gene Symbol: LOC4328604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK120869.1 | - | 3 - 4 | 1191 | 85 | |
AK121281.1 | - | 3 - 4 | 1159 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
