Assay Details
Gene Symbol: LOC4329657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772155.1 | XP_015627641.1 | 6 - 7 | 970 | 84 | |
GenBank mRNA | AK108057.1 | - | 6 - 7 | 970 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
