Assay Details
| Gene Symbol: LOC4329812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768564.2 | XP_015624050.1 | 1 - 2 | 188 | 101 | |
| GenBank mRNA | AK063583.1 | - | 1 - 2 | 188 | 101 | |
| AK105378.1 | - | 1 - 2 | 189 | 101 | ||
More Information
Set Membership: |
|

LOC4329812
Genomic Map