Assay Details
| Gene Symbol: LOC4329902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770451.2 | XP_015625937.1 | 1 - 2 | 1090 | 111 | |
| GenBank mRNA | AK064340.1 | - | 1 - 2 | 1070 | 111 | |
| AK073602.1 | - | 2 - 3 | 741 | 111 | ||
More Information
Set Membership: |
|

LOC4329902
Genomic Map