Assay Details
| Gene Symbol: LOC4329937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769839.2 | XP_015625325.1 | 2 - 3 | 870 | 100 | |
| GenBank mRNA | AK071922.1 | - | 2 - 3 | 338 | 100 | |
| AK099756.1 | - | 2 - 3 | 737 | 100 | ||
More Information
Set Membership: |
|

LOC4329937
Genomic Map