Assay Details
| Gene Symbol: LOC4329937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769839.2 | XP_015625325.1 | 9 - 10 | 1528 | 96 | |
| GenBank mRNA | AK071922.1 | - | 9 - 10 | 996 | 96 | |
| AK099756.1 | - | 9 - 10 | 1395 | 96 | ||
More Information
Set Membership: |
|

LOC4329937
Genomic Map