Assay Details
Gene Symbol: LOC4330086 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767559.2 | XP_015623045.1 | 1 - 2 | 314 | 78 | |
GenBank mRNA | AK069779.1 | - | 1 - 2 | 260 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
