Assay Details
| Gene Symbol: LOC4330235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767260.1 | XP_015622746.1 | 2 - 3 | 386 | 66 | |
| GenBank mRNA | AK058456.1 | - | 2 - 3 | 336 | 66 | |
| AK105165.1 | - | 2 - 3 | 329 | 66 | ||
More Information
Set Membership: |
|

LOC4330235
Genomic Map