Assay Details
| Gene Symbol: LOC4330235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767260.1 | XP_015622746.1 | 5 - 6 | 730 | 73 | |
| GenBank mRNA | AK058456.1 | - | 5 - 6 | 680 | 73 | |
| AK105165.1 | - | 5 - 6 | 673 | 73 | ||
More Information
Set Membership: |
|

LOC4330235
Genomic Map