Assay Details
Gene Symbol: LOC4330314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769445.2 | XP_015624931.1 | 2 - 3 | 489 | 80 | |
GenBank mRNA | AK108170.1 | - | 2 - 3 | 467 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
