Assay Details
| Gene Symbol: LOC4330389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770607.2 | XP_015626093.1 | 1 - 2 | 391 | 62 | |
| GenBank mRNA | AK103651.1 | - | 1 - 2 | 243 | 62 | |
More Information
Set Membership: |
|

LOC4330389
Genomic Map