Assay Details
Gene Symbol: LOC4330406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771062.2 | XP_015626548.1 | 3 - 4 | 869 | 83 | |
XM_015771063.2 | XP_015626549.1 | 3 - 4 | 882 | 83 | ||
XM_015771064.2 | XP_015626550.1 | 2 - 3 | 785 | 83 | ||
GenBank mRNA | AJ495787.1 | - | 3 - 4 | 779 | 83 | |
AK066018.1 | - | 3 - 4 | 780 | 83 | ||
EU837251.1 | - | 3 - 4 | 708 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
