Assay Details
Gene Symbol: LOC4330406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771062.2 | XP_015626548.1 | 4 - 5 | 1109 | 96 | |
XM_015771063.2 | XP_015626549.1 | 4 - 5 | 1122 | 96 | ||
XM_015771064.2 | XP_015626550.1 | 3 - 4 | 1025 | 96 | ||
GenBank mRNA | AJ495787.1 | - | 4 - 5 | 1019 | 96 | |
AK066018.1 | - | 4 - 5 | 1020 | 96 | ||
EU837251.1 | - | 4 - 5 | 948 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
