Assay Details
| Gene Symbol: LOC4330561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771775.2 | XP_015627261.1 | 3 - 4 | 696 | 74 | |
| XM_015771776.2 | XP_015627262.1 | 3 - 4 | 695 | 74 | ||
| XM_015771777.2 | XP_015627263.1 | 3 - 4 | 696 | 74 | ||
| GenBank mRNA | AK100043.1 | - | 3 - 4 | 668 | 74 | |
| EU605819.1 | - | 1 - 2 | 317 | 74 | ||
More Information
Set Membership: |
|

LOC4330561
Genomic Map