Assay Details
| Gene Symbol: LOC4330561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771775.2 | XP_015627261.1 | 7 - 8 | 1250 | 95 | |
| XM_015771776.2 | XP_015627262.1 | 7 - 8 | 1249 | 95 | ||
| XM_015771777.2 | XP_015627263.1 | 7 - 8 | 1250 | 95 | ||
| GenBank mRNA | AK100043.1 | - | 7 - 8 | 1222 | 95 | |
| EU605819.1 | - | 5 - 6 | 871 | 95 | ||
More Information
Set Membership: |
|

LOC4330561
Genomic Map