Assay Details
| Gene Symbol: LOC4330561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771775.2 | XP_015627261.1 | 8 - 9 | 1295 | 107 | |
| XM_015771776.2 | XP_015627262.1 | 8 - 9 | 1294 | 107 | ||
| XM_015771777.2 | XP_015627263.1 | 8 - 9 | 1295 | 107 | ||
| GenBank mRNA | AK100043.1 | - | 8 - 9 | 1267 | 107 | |
| EU605819.1 | - | 6 - 7 | 916 | 107 | ||
More Information
Set Membership: |
|

LOC4330561
Genomic Map