Assay Details
| Gene Symbol: LOC4330573 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767674.2 | XP_015623160.1 | 8 - 9 | 681 | 91 | |
| XM_015767675.1 | XP_015623161.1 | 8 - 9 | 681 | 91 | ||
| GenBank mRNA | AK073256.1 | - | 7 - 8 | 534 | 91 | |
| AK105251.1 | - | 1489 | 91 | |||
More Information
Set Membership: |
|

LOC4330573
Genomic Map