Assay Details
| Gene Symbol: LOC4330573 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767674.2 | XP_015623160.1 | 10 - 11 | 837 | 135 | |
| GenBank mRNA | AK073256.1 | - | 9 - 10 | 690 | 135 | |
| AK105251.1 | - | 1645 | 135 | |||
More Information
Set Membership: |
|

LOC4330573
Genomic Map