Assay Details
| Gene Symbol: LOC4330749 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772108.2 | XP_015627594.1 | 4 - 5 | 552 | 72 | |
| XM_015772109.2 | XP_015627595.1 | 4 - 5 | 554 | 72 | ||
| GenBank mRNA | AK062267.1 | - | 4 - 5 | 485 | 72 | |
| AK066184.1 | - | 4 - 5 | 489 | 72 | ||
More Information
Set Membership: |
|

LOC4330749
Genomic Map