Assay Details
| Gene Symbol: LOC4330868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772116.2 | XP_015627602.1 | 7 - 8 | 1490 | 68 | |
| GenBank mRNA | AB040053.1 | - | 7 - 8 | 1307 | 68 | |
| AK102740.1 | - | 7 - 8 | 1497 | 68 | ||
| AK111614.1 | - | 7 - 8 | 1316 | 68 | ||
| AK112098.1 | - | 8 - 9 | 1014 | 68 | ||
More Information
Set Membership: |
|

LOC4330868
Genomic Map