Assay Details
| Gene Symbol: LOC4330921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768091.2 | XP_015623577.1 | 5 - 6 | 718 | 71 | |
| GenBank mRNA | AK067608.1 | - | 5 - 6 | 676 | 71 | |
More Information
Set Membership: |
|

LOC4330921
Genomic Map