Assay Details
Gene Symbol: LOC4331069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770166.1 | XP_015625652.1 | 3 - 4 | 453 | 64 | |
GenBank mRNA | AK073740.1 | - | 3 - 4 | 459 | 64 |
Gene Symbol: LOC4340405 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK065642.1 | - | 243 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
